Product Details

SNP ID
rs4830982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15682840 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTACCCCAGCAGAAGGCTAGTGTAC[A/G]CTTGTAACCAAGAATAACCCCAGAT
Phenotype
MIM: 300631
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CA5BP1 PubMed Links

Gene Details

Gene
CA5BP1
Gene Name
carbonic anhydrase 5B pseudogene 1
There are no transcripts associated with this gene.

Gene
TMEM27
Gene Name
transmembrane protein 27
There are no transcripts associated with this gene.

View Full Product Details