Product Details

SNP ID
rs537938359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:93896632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAACAACCCCTGCAAACTGCTC[C/T]GCCAAGTAAAGTAATTCATACTTTG
Phenotype
MIM: 176880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PROS1 PubMed Links

Gene Details

Gene
PROS1
Gene Name
protein S (alpha)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000313.3 1337 Silent Mutation NP_000304.2
NM_001314077.1 1337 Silent Mutation NP_001301006.1

View Full Product Details