Product Details
- SNP ID
-
rs4377234
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:41482362 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGAAAGACAGTGGTGGAAGTGAA[A/G]AAGAAGGGCAAATGGGACCTCCCCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KC6
PubMed Links
Gene Details
- Gene
- KC6
- Gene Name
- keratoconus gene 6
There are no transcripts associated with this gene.
View Full Product Details