Product Details
- SNP ID
-
rs4773145
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:110310684 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAATTGTGGCAAGGACTCTTTCCTG[C/T]GTAGGTCAGTTCAGGAAAAAGGGCT
- Phenotype
-
MIM: 120130
MIM: 120090
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COL4A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115454213] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COL4A1
- Gene Name
- collagen type IV alpha 1 chain
There are no transcripts associated with this gene.
- Gene
- COL4A2
- Gene Name
- collagen type IV alpha 2
There are no transcripts associated with this gene.
View Full Product Details