Product Details

SNP ID
rs4903545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77132148 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCCCTACTCCCTTATATGCCCCAG[A/G]GGTCTGGTCCTCTAGGTTTCAGAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZDHHC22 PubMed Links

Gene Details

Gene
ZDHHC22
Gene Name
zinc finger DHHC-type containing 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174976.2 2530 UTR 3 NP_777636.2
XM_006720119.3 2530 UTR 3 XP_006720182.1
XM_011536661.2 2530 UTR 3 XP_011534963.1

View Full Product Details