Product Details

SNP ID
rs4878204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:38409477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAATCCACAGATGGCTCTCAGTG[C/G]TAGTGAGCTCTGTATCCCTGGAGGT
Phenotype
MIM: 610413
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IGFBPL1 PubMed Links

Gene Details

Gene
IGFBPL1
Gene Name
insulin like growth factor binding protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007563.2 Intron NP_001007564.1
XM_017014699.1 Intron XP_016870188.1

View Full Product Details