Product Details

SNP ID
rs4565916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57488103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAGACTTATGTCCTGACAGGTG[A/G]CTGCAGCCCCTGTGTTAAAAATAGA
Phenotype
MIM: 603733
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC43A1 PubMed Links

Gene Details

Gene
SLC43A1
Gene Name
solute carrier family 43 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198810.1 Intron NP_001185739.1
NM_003627.5 Intron NP_003618.1

View Full Product Details