Product Details

SNP ID
rs4720144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:33908851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAATTATGGGGTCCAAGTTGTCA[C/T]AGTCATTGCTGCTTCCCAAACGTTT
Phenotype
MIM: 608699
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BMPER PubMed Links

Gene Details

Gene
BMPER
Gene Name
BMP binding endothelial regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133468.4 Intron NP_597725.1

View Full Product Details