Product Details
- SNP ID
-
rs4401853
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:51817961 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAACACCCAAATTAGATGATACTTA[C/T]TGTTTTAACTAGCTATAAAATTCCA
- Phenotype
-
MIM: 615904
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PCMTD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs199619900] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PCMTD1
- Gene Name
- protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1
- Gene
- PXDNL
- Gene Name
- peroxidasin like
There are no transcripts associated with this gene.
View Full Product Details