Product Details

SNP ID
rs4546189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:79828819 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCAGCTTCTCCTTATTAACTTTG[C/T]GAATGGATCCATCACATCAGAATCA
Phenotype
MIM: 609717
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCAT4 PubMed Links

Gene Details

Gene
PCAT4
Gene Name
prostate cancer associated transcript 4 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details