Product Details

SNP ID
rs4839536
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:110603099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATAAGCCGGTGATGAGGATGTGC[A/C]TGAAAGCCATGATAGATATTCTGTG
Phenotype
MIM: 176262
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNA2 PubMed Links

Gene Details

Gene
KCNA2
Gene Name
potassium voltage-gated channel subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204269.1 Intron NP_001191198.1
NM_004974.3 Intron NP_004965.1

View Full Product Details