Product Details
- SNP ID
-
rs4729090
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:93428291 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAGGGCACTACTATCTTCACTTTA[C/T]GGATGGGGCACTGAGGTTATGCAAT
- Phenotype
-
MIM: 114131
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CALCR
PubMed Links
Gene Details
- Gene
- CALCR
- Gene Name
- calcitonin receptor
View Full Product Details