Product Details

SNP ID
rs4347635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23296612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGGAAATATTGGAGACAGAAA[A/G]AGGGATAACAAAAAGGACATGGATC
Phenotype
MIM: 600760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCNN1B PubMed Links

Gene Details

Gene
SCNN1B
Gene Name
sodium channel epithelial 1 beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000336.2 Intron NP_000327.2

View Full Product Details