Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001145168.1 | Intron | NP_001138640.1 | |||
NM_001145169.1 | Intron | NP_001138641.1 | |||
NM_001321971.1 | Intron | NP_001308900.1 | |||
NM_001321975.1 | Intron | NP_001308904.1 | |||
NM_153835.3 | Intron | NP_722577.2 | |||
XM_011532621.2 | Intron | XP_011530923.1 | |||
XM_011532622.2 | Intron | XP_011530924.1 | |||
XM_011532623.2 | Intron | XP_011530925.1 | |||
XM_017003505.1 | Intron | XP_016858994.1 | |||
XM_017003506.1 | Intron | XP_016858995.1 |