Product Details

SNP ID
rs4821626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37284086 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCAGTCAGCAGCAGAGCAGGGAC[A/G]AGAACCTGCGGGAGAGAGCCCCAGG
Phenotype
MIM: 606514
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYTH4 PubMed Links

Gene Details

Gene
CYTH4
Gene Name
cytohesin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318024.1 69 Intron NP_001304953.1
NM_013385.4 69 Intron NP_037517.1
XM_011530147.2 69 UTR 5 XP_011528449.1
XM_011530148.1 69 Intron XP_011528450.1

View Full Product Details