Product Details
- SNP ID
-
rs4714773
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:44274791 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCAGAGGGGCAGTTCATGGCCAGG[A/G]CCTGACCCCTAGAGGAAGCAGGGCT
- Phenotype
-
MIM: 604548
MIM: 186975
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NFKBIE
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115381229] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NFKBIE
- Gene Name
- NFKB inhibitor epsilon
There are no transcripts associated with this gene.
- Gene
- TCTE1
- Gene Name
- t-complex-associated-testis-expressed 1
There are no transcripts associated with this gene.
- Gene
- TMEM151B
- Gene Name
- transmembrane protein 151B
View Full Product Details