Product Details

SNP ID
rs4292863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:41085912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCGGCGAGGAGCGAGTTCCGGC[C/G]CCGGTGTGCAGCCTTTTGGTTGAGA
Phenotype
MIM: 300072
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
USP9X PubMed Links

Gene Details

Gene
USP9X
Gene Name
ubiquitin specific peptidase 9, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039590.2 278 UTR 5 NP_001034679.2
NM_001039591.2 278 UTR 5 NP_001034680.2

View Full Product Details