Product Details

SNP ID
rs4903320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75434117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGTTCCTTCTGGCTGTTACTGG[A/T]TCCTTGGTGCTGCCACCTCCACTGC
Phenotype
MIM: 608657
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
JDP2 PubMed Links

Gene Details

Gene
JDP2
Gene Name
Jun dimerization protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135047.1 Intron NP_001128519.1
NM_001135048.1 Intron NP_001128520.1
NM_001135049.1 Intron NP_001128521.1
NM_130469.3 Intron NP_569736.1
Gene
LOC101928377
Gene Name
uncharacterized LOC101928377
There are no transcripts associated with this gene.

View Full Product Details