Product Details

SNP ID
rs4660233
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:42848230 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTGGAGGCTGAATAGCTCATTGC[A/G]TAGAGATACTGTAGCTGTAGGTCAG
Phenotype
MIM: 609017
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERMAP PubMed Links

Gene Details

Gene
ERMAP
Gene Name
erythroblast membrane associated protein (Scianna blood group)
There are no transcripts associated with this gene.

Gene
LOC339539
Gene Name
uncharacterized LOC339539
There are no transcripts associated with this gene.

Gene
ZNF691
Gene Name
zinc finger protein 691
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242739.1 1555 Intron NP_001229668.1
NM_015911.3 1555 Intron NP_056995.1
XM_006710671.3 1555 Intron XP_006710734.1
XM_017001400.1 1555 Intron XP_016856889.1
XM_017001401.1 1555 UTR 5 XP_016856890.1
XM_017001402.1 1555 Intron XP_016856891.1
XM_017001403.1 1555 UTR 5 XP_016856892.1

View Full Product Details