Product Details

SNP ID
rs4319915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:11540907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTTGAAAGTGAGACAGGTTGAG[G/T]ACGCAGTCCGGAGAGTGTTGCTTTT
Phenotype
MIM: 611736
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GREB1 PubMed Links

Gene Details

Gene
GREB1
Gene Name
growth regulation by estrogen in breast cancer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014668.3 Intron NP_055483.2
NM_033090.2 Intron NP_149081.1
NM_148903.2 Intron NP_683701.2
Gene
MIR4429
Gene Name
microRNA 4429
There are no transcripts associated with this gene.

View Full Product Details