Product Details

SNP ID
hCV27998847
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28259743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTCCTCTCCCACAGTGAGCATGT[A/C]CCTCCCCTGCTCAGAGCCCTCCATG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SBK1 PubMed Links

Gene Details

Gene
SBK1
Gene Name
SH3 domain binding kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024401.2 Intron NP_001019572.1

View Full Product Details