Product Details

SNP ID
rs4885028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:72708597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATCTACAAAGTCATAAATAATAC[C/T]ATCAACTCTTATGGCCTGGAAGTAT
Phenotype
MIM: 613448
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MZT1 PubMed Links

Gene Details

Gene
MZT1
Gene Name
mitotic spindle organizing protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001071775.2 2172 UTR 3 NP_001065243.1

View Full Product Details