Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001012426.1 | Intron | NP_001012426.1 | |||
NM_001012427.1 | Intron | NP_001012427.1 | |||
NM_138457.2 | Intron | NP_612466.1 | |||
XM_006714991.3 | Intron | XP_006715054.1 | |||
XM_011514289.2 | Intron | XP_011512591.1 | |||
XM_011514290.2 | Intron | XP_011512592.1 | |||
XM_011514291.2 | Intron | XP_011512593.1 | |||
XM_011514292.2 | Intron | XP_011512594.1 | |||
XM_011514293.2 | Intron | XP_011512595.1 | |||
XM_017010233.1 | Intron | XP_016865722.1 | |||
XM_017010234.1 | Intron | XP_016865723.1 |