Product Details

SNP ID
rs4785217
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:50555332 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACTGTCTGTGGAAAGCCTCGTGC[A/G]TTGGTGGCTCTTGGCACATCTGAGT
Phenotype
MIM: 607851
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NKD1 PubMed Links

Gene Details

Gene
NKD1
Gene Name
naked cuticle homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033119.4 Intron NP_149110.1
XM_011523399.2 Intron XP_011521701.1
XM_011523400.2 Intron XP_011521702.1
XM_017023804.1 Intron XP_016879293.1

View Full Product Details