Product Details
- SNP ID
-
rs4686786
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:186538560 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGTGGCGAGCACTGTATTGCTGG[C/T]CCCTAGAAGCATTTAAGGAGAGGCA
- Phenotype
-
MIM: 123730
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CRYGS
PubMed Links
Gene Details
- Gene
- CRYGS
- Gene Name
- crystallin gamma S
There are no transcripts associated with this gene.
- Gene
- TBCCD1
- Gene Name
- TBCC domain containing 1
There are no transcripts associated with this gene.
View Full Product Details