Product Details

SNP ID
hCV28012607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:37744495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGTGTCTGTTGTAGCACCTTGG[A/T]AAAGGAGAGCTGCCGTGTCTGAGGG
Phenotype
MIM: 611605
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ERLIN2 PubMed Links

Gene Details

Gene
ERLIN2
Gene Name
ER lipid raft associated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003790.3 Intron NP_001003790.1
NM_001003791.2 Intron NP_001003791.1
NM_007175.6 Intron NP_009106.1
Gene
LOC102723701
Gene Name
uncharacterized LOC102723701
There are no transcripts associated with this gene.

Gene
LOC728024
Gene Name
chromosome X open reading frame 56 pseudogene
There are no transcripts associated with this gene.

View Full Product Details