Product Details
- SNP ID
-
rs4623981
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52839292 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGTGCAGCAGGACCCAGGGCTGC[C/T]TTTCCCACTACCGAGTCCACAAGTG
- Phenotype
-
MIM: 611159
MIM: 611160
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT78
PubMed Links
Gene Details
- Gene
- KRT78
- Gene Name
- keratin 78
- Gene
- KRT79
- Gene Name
- keratin 79
There are no transcripts associated with this gene.
View Full Product Details