Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001316957.1 | Intron | NP_001303886.1 | |||
NM_001316958.1 | Intron | NP_001303887.1 | |||
NM_001316960.1 | Intron | NP_001303889.1 | |||
NM_001316961.1 | Intron | NP_001303890.1 | |||
NM_001316962.1 | Intron | NP_001303891.1 | |||
NM_024684.3 | Intron | NP_078960.1 | |||
XM_011544968.2 | Intron | XP_011543270.1 | |||
XM_011544969.2 | Intron | XP_011543271.1 | |||
XM_017017620.1 | Intron | XP_016873109.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_016578.3 | Intron | NP_057662.3 | |||
XM_005274051.2 | Intron | XP_005274108.1 | |||
XM_017017923.1 | Intron | XP_016873412.1 |