Product Details

SNP ID
rs4806701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54071325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTTCCTTGATATTCACTCCAC[C/T]TTAGCTCTCTTCCTTCGTTTATTTG
Phenotype
MIM: 616802 MIM: 616804
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TARM1 PubMed Links

Gene Details

Gene
TARM1
Gene Name
T cell-interacting, activating receptor on myeloid cells 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135686.2 Intron NP_001129158.2
XM_005278278.3 Intron XP_005278335.1
Gene
VSTM1
Gene Name
V-set and transmembrane domain containing 1
There are no transcripts associated with this gene.

View Full Product Details