Product Details
- SNP ID
-
rs4434550
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:88763052 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTTGACATAATAAGGATTTCTT[C/T]TTTTGTAGTTCTGATATTATGGATA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF804B
PubMed Links
Gene Details
- Gene
- ZNF804B
- Gene Name
- zinc finger protein 804B
There are no transcripts associated with this gene.
View Full Product Details