Product Details

SNP ID
rs4567686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:98651452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGGTGGTGGCTGGGGGGACAAAG[A/G]AGGGTTTTTATCTTTAAAAGATAAG
Phenotype
MIM: 147370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGF1R PubMed Links

Gene Details

Gene
IGF1R
Gene Name
insulin like growth factor 1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000875.4 Intron NP_000866.1
NM_001291858.1 Intron NP_001278787.1
Gene
IRAIN
Gene Name
IGF1R antisense imprinted non-protein coding RNA
There are no transcripts associated with this gene.

View Full Product Details