Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001164458.1 | 4622 | Intron | NP_001157930.1 | ||
NM_001164459.1 | 4622 | Intron | NP_001157931.1 | ||
XM_005250043.4 | 4622 | Intron | XP_005250100.3 | ||
XM_011516502.2 | 4622 | Intron | XP_011514804.1 | ||
XM_011516506.2 | 4622 | Intron | XP_011514808.1 | ||
XM_011516507.2 | 4622 | Intron | XP_011514809.1 | ||
XM_011516508.2 | 4622 | Intron | XP_011514810.1 | ||
XM_011516511.2 | 4622 | Intron | XP_011514813.1 | ||
XM_017012548.1 | 4622 | UTR 3 | XP_016868037.1 | ||
XM_017012549.1 | 4622 | Intron | XP_016868038.1 | ||
XM_017012550.1 | 4622 | UTR 3 | XP_016868039.1 | ||
XM_017012551.1 | 4622 | UTR 3 | XP_016868040.1 | ||
XM_017012552.1 | 4622 | Intron | XP_016868041.1 | ||
XM_017012553.1 | 4622 | Intron | XP_016868042.1 |