Product Details

SNP ID
rs4878627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35682552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGCAGTGAAAGCCCCATGTTG[C/G]TGATATCCAACATTTTTCCAAGCTC
Phenotype
MIM: 603179 MIM: 190990
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CA9 PubMed Links

Gene Details

Gene
CA9
Gene Name
carbonic anhydrase 9
There are no transcripts associated with this gene.

Gene
TPM2
Gene Name
tropomyosin 2 (beta)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301226.1 Intron NP_001288155.1
NM_001301227.1 Intron NP_001288156.1
NM_003289.3 Intron NP_003280.2
NM_213674.1 Intron NP_998839.1

View Full Product Details