Product Details

SNP ID
rs4844286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71151724 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTTCAGAGGGATTCACAAGGGAC[G/T]TATCTTCTAAGTCAGGGATGATGGG
Phenotype
MIM: 300188 MIM: 300336
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MED12 PubMed Links

Gene Details

Gene
MED12
Gene Name
mediator complex subunit 12
There are no transcripts associated with this gene.

Gene
NLGN3
Gene Name
neuroligin 3
There are no transcripts associated with this gene.

View Full Product Details