Product Details
- SNP ID
-
rs4743940
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:93953447 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCCCAGGCCTCAGCTGCCTCTGA[A/G]ATCTCGGGGTTCCTGATGAACATGT
- Phenotype
-
MIM: 603260
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BARX1
PubMed Links
Gene Details
- Gene
- BARX1
- Gene Name
- BARX homeobox 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021570.3 |
|
Intron |
|
|
NP_067545.3 |
View Full Product Details