Product Details

SNP ID
rs7118544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46607493 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAAAGAGAATAAAATAAGTAAGA[A/T]GAATACTGAAAACTGGAGATGCTAA
Phenotype
MIM: 615088 MIM: 615086
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATG13 PubMed Links

Gene Details

Gene
ATG13
Gene Name
autophagy related 13
There are no transcripts associated with this gene.

Gene
HARBI1
Gene Name
harbinger transposase derived 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173811.3 Intron NP_776172.1
XM_011520025.1 Intron XP_011518327.1
XM_011520026.2 Intron XP_011518328.1
XM_011520027.1 Intron XP_011518329.1
XM_011520028.2 Intron XP_011518330.1
XM_011520029.2 Intron XP_011518331.1
XM_017017602.1 Intron XP_016873091.1
XM_017017603.1 Intron XP_016873092.1
XM_017017604.1 Intron XP_016873093.1
XM_017017605.1 Intron XP_016873094.1
XM_017017606.1 Intron XP_016873095.1

View Full Product Details