Product Details
- SNP ID
-
rs6581561
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:64411509 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTTTTTCAGAAAATTAAAACCAAT[A/G]CATTATTAGTGGTTTTAATACAGAA
- Phenotype
-
MIM: 603180
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
XPOT
PubMed Links
Gene Details
- Gene
- XPOT
- Gene Name
- exportin for tRNA
View Full Product Details