Product Details

SNP ID
rs6426124
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:224437079 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCATTTATTAGGCTCCTCAATCAC[A/G]AAGGCTGCTTGTGATGTAACTTGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNIH3 PubMed Links

Gene Details

Gene
CNIH3
Gene Name
cornichon family AMPA receptor auxiliary protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322302.1 Intron NP_001309231.1
NM_001322303.1 Intron NP_001309232.1
NM_001322304.1 Intron NP_001309233.1
NM_001322305.1 Intron NP_001309234.1
NM_152495.1 Intron NP_689708.1
Gene
WDR26
Gene Name
WD repeat domain 26
There are no transcripts associated with this gene.

View Full Product Details