Product Details
- SNP ID
-
rs7980646
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:108912099 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAAAGAAGAAAGCCTGGGGCTGTG[A/G]GTGTGGGAGAAACCTACTGAACAGG
- Phenotype
-
MIM: 611699
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SVOP
PubMed Links
Gene Details
- Gene
- SVOP
- Gene Name
- SV2 related protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018711.4 |
2455 |
UTR 3 |
|
|
NP_061181.1 |
View Full Product Details