Product Details

SNP ID
rs7815323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:69670668 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATACCTGGCACCTGATCAGTGATC[A/G]CCAAACTGGATGACTGTTCATGGCA
Phenotype
MIM: 613543 MIM: 610012
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLCO5A1 PubMed Links

Gene Details

Gene
SLCO5A1
Gene Name
solute carrier organic anion transporter family member 5A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146008.1 5290 Intron NP_001139480.1
NM_001146009.1 5290 Intron NP_001139481.1
NM_030958.2 5290 Intron NP_112220.2
XM_005251313.2 5290 UTR 3 XP_005251370.1
XM_017013883.1 5290 UTR 3 XP_016869372.1
XM_017013884.1 5290 Intron XP_016869373.1
XM_017013885.1 5290 UTR 3 XP_016869374.1
XM_017013886.1 5290 Intron XP_016869375.1
Gene
SULF1
Gene Name
sulfatase 1
There are no transcripts associated with this gene.

View Full Product Details