Product Details

SNP ID
rs7222985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38363338 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCTTTCTAGAACTTCCACTGATG[A/G]CTTTTGTTTATTTTTGACACAGGGC
Phenotype
MIM: 608788
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SOCS7 PubMed Links

Gene Details

Gene
SOCS7
Gene Name
suppressor of cytokine signaling 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014598.3 Intron NP_055413.1
XM_017024551.1 Intron XP_016880040.1
XM_017024552.1 Intron XP_016880041.1

View Full Product Details