Product Details

SNP ID
rs4889605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30989774 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGCTCAGGGCGTGGCAAGAGCGTG[C/T]GTCGACCCACGGGTACATGGGATGG
Phenotype
MIM: 607764 MIM: 611052 MIM: 601485
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSD3B7 PubMed Links

Gene Details

Gene
HSD3B7
Gene Name
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142777.1 4156 Intron NP_001136249.1
NM_001142778.1 4156 Intron NP_001136250.1
NM_025193.3 4156 Intron NP_079469.2
XM_005255601.3 4156 Intron XP_005255658.2
XM_011545960.2 4156 Intron XP_011544262.1
XM_011545961.1 4156 Intron XP_011544263.1
XM_011545962.2 4156 Intron XP_011544264.1
XM_017023732.1 4156 Intron XP_016879221.1
Gene
SETD1A
Gene Name
SET domain containing 1A
There are no transcripts associated with this gene.

Gene
STX1B
Gene Name
syntaxin 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052874.4 4156 UTR 3 NP_443106.1
XM_017022893.1 4156 UTR 3 XP_016878382.1

View Full Product Details