Product Details

SNP ID
rs7624120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:64691594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTATGTACCAGACCATGTGTACT[A/C]TGTCCTCTCTGATCCGGTTTCCTAT
Phenotype
MIM: 605421
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTS9 PubMed Links

Gene Details

Gene
ADAMTS9
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 9
There are no transcripts associated with this gene.

Gene
ADAMTS9-AS2
Gene Name
ADAMTS9 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details