Product Details

SNP ID
rs6540961
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:9590671 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAAGTCCTTTCACGCCCCTGACG[A/G]CACCAGCAGGTCACGGTGTGGCATT
Phenotype
MIM: 610816
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A33 PubMed Links

Gene Details

Gene
SLC25A33
Gene Name
solute carrier family 25 member 33
There are no transcripts associated with this gene.

Gene
TMEM201
Gene Name
transmembrane protein 201
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010866.3 Intron NP_001010866.1
NM_001130924.2 Intron NP_001124396.2
XM_006710417.3 Intron XP_006710480.1
XM_011540910.2 Intron XP_011539212.1
XM_017000549.1 Intron XP_016856038.1
XM_017000550.1 Intron XP_016856039.1
XM_017000551.1 Intron XP_016856040.1
XM_017000552.1 Intron XP_016856041.1

View Full Product Details