Product Details

SNP ID
rs7586642
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:191949903 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAACCTAGCCACTGAGTCCTGTAC[A/G]AACTAATGTGCTGTCTCAAGATGAG
Phenotype
MIM: 605734
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEFF2 PubMed Links

Gene Details

Gene
TMEFF2
Gene Name
transmembrane protein with EGF like and two follistatin like domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305134.1 1979 UTR 3 NP_001292063.1
NM_001305145.1 1979 Intron NP_001292074.1
NM_016192.3 1979 UTR 3 NP_057276.2
XM_011510890.2 1979 Intron XP_011509192.1
XM_017003739.1 1979 UTR 3 XP_016859228.1
XM_017003740.1 1979 Intron XP_016859229.1

View Full Product Details