Product Details
- SNP ID
-
rs6866492
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:40916026 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGAGTTATAAATTGGATGCATTTG[A/G]TATTCTTAGAGAATCAGTAAAGATT
- Phenotype
-
MIM: 217070
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C7
PubMed Links
Gene Details
- Gene
- C7
- Gene Name
- complement component 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000587.2 |
|
Intron |
|
|
NP_000578.2 |
View Full Product Details