Product Details

SNP ID
rs7171160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:69288177 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTGGAGGCTCTAGGGAATGGTCC[C/G]TTTCCTTGCCTTCTCCAACTTCCAG
Phenotype
MIM: 607781
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PAQR5 PubMed Links

Gene Details

Gene
PAQR5
Gene Name
progestin and adipoQ receptor family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104554.1 Intron NP_001098024.1
NM_017705.3 Intron NP_060175.3
XM_005254495.4 Intron XP_005254552.1
XM_011521720.2 Intron XP_011520022.1
XM_011521722.2 Intron XP_011520024.1
XM_011521723.2 Intron XP_011520025.1
XM_011521725.2 Intron XP_011520027.1
XM_011521727.2 Intron XP_011520029.1
XM_017022360.1 Intron XP_016877849.1
XM_017022361.1 Intron XP_016877850.1
XM_017022362.1 Intron XP_016877851.1
XM_017022363.1 Intron XP_016877852.1
XM_017022364.1 Intron XP_016877853.1
XM_017022365.1 Intron XP_016877854.1
XM_017022366.1 Intron XP_016877855.1
XM_017022367.1 Intron XP_016877856.1
XM_017022368.1 Intron XP_016877857.1

View Full Product Details