Product Details

SNP ID
rs6494406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63511590 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTATAGGCCGAAATTCATATTGC[A/T]TTAGTTCAGACATAATAAACCAGCC
Phenotype
MIM: 604728
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
USP3 PubMed Links

Gene Details

Gene
USP3
Gene Name
ubiquitin specific peptidase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256702.1 Intron NP_001243631.1
NM_006537.3 Intron NP_006528.2
XM_017022763.1 Intron XP_016878252.1
XM_017022764.1 Intron XP_016878253.1
XM_017022765.1 Intron XP_016878254.1
XM_017022766.1 Intron XP_016878255.1
XM_017022767.1 Intron XP_016878256.1
XM_017022768.1 Intron XP_016878257.1

View Full Product Details