Product Details
- SNP ID
-
rs7716523
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:150489923 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGTCTTGTCCTTCAGGGTTCCTGA[A/G]GATGGAGAAAGCAACAAGGGTGACA
- Phenotype
-
MIM: 600853
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC102546298
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111288376] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC102546298
- Gene Name
- uncharacterized LOC102546298
There are no transcripts associated with this gene.
- Gene
- NDST1
- Gene Name
- N-deacetylase and N-sulfotransferase 1
View Full Product Details