Product Details

SNP ID
rs6495760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:35218650 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTTTATTTAACATATGATAATCC[G/T]CTCTTCTTTGGACTCTCTTATTTAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DPH6 PubMed Links

Gene Details

Gene
DPH6
Gene Name
diphthamine biosynthesis 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001141972.1 2764 Intron NP_001135444.1
NM_080650.3 2764 Intron NP_542381.1
XM_011522160.1 2764 Intron XP_011520462.1
XM_017022707.1 2764 Intron XP_016878196.1
XM_017022708.1 2764 UTR 3 XP_016878197.1
XM_017022709.1 2764 Intron XP_016878198.1

View Full Product Details